Brady® Safety Inspection Timers, Brady Worldwide
Supplier: Brady Worldwide
150740 150741 150743 150744
76360-006PK 512.66 CAD
76360-006 76360-008 76360-010 76360-012 76360-014
Brady® Safety Inspection Timers, Brady Worldwide
Timers Digital Timers
Shed some light on inspections by supplementing inspection tags with the inspection timer, and get an inspection status at a glance.
  • Review and reset; after completing an inspection, reset the timer so it is ready for the next inspection
  • Perfect for facilities that have internal audit and inspection schedules
  • Sturdy ABS plastic construction
Unlike tags, the bright color LED lights provide an inspection status from a distance and break through the grime of any facility
IP 65 rated timers will survive harsh industrial environments, including exposure to water, dust, extreme temperatures and more.
Safety inspection timers are ideal for use in facilities that have multiple eyewash stations, emergency showers, emergency exit doors, exit signs, fire extinguishers, or first aid kits.
Certifications: Ingress Protection Rating IP 65.
Delivery information: All timers are supplied with three AAA Alkaline batteries.