Brady® BatteryBlock™ Power Connector Lockout - Forklift, Brady Worldwide
Supplier: Brady Worldwide
150841
76359-914EA 56.82 CAD
76359-914
Brady® BatteryBlock™ Power Connector Lockout - Forklift, Brady Worldwide
Security Locks
For forklift safety, proper operation is a main concern, but a secure lockout can be just as important. Keep forklifts and lift vehicles locked out and locked down with the BatteryBlock™ power connector lockout.
  • Device fits on 175 A and 350 A power connectors with and without handles
  • Single-piece design is easy to store and manage so users don't have to worry about losing pieces
  • Manage use and safety by applying two padlocks from authorized employees
Non-conductive pins ensures the device stays secure during maintenance projects and off-shift periods.
Certifications: OSHA 29 CFR 1910.147.