Brady® BatteryBlock™ Battery Cable Lockout - Commercial Vehicles, Brady Worldwide
Supplier: Brady Worldwide
150820 150821 150822 150823
76359-906EA 41.08 CAD
76359-906 76359-908 76359-910 76359-912
Brady® BatteryBlock™ Battery Cable Lockout - Commercial Vehicles, Brady Worldwide
Security Locks
Get easy prevention of an accidental battery connection during service projects on commercial vehicles with the BatteryBlock™ Battery Cable Lockout.
  • A tool that fits virtually any space
  • Low profile design means it easily fits into tight working areas
  • Small size fits endline battery cables
  • Keep it locked out with space for two safety padlocks (sold separetely)
Innovative non-conductive post slips through the terminal end of the battery cable to prevent it from slipping or being pulled out of the device.
Certifications: OSHA 29 CFR 1910.147.
Order Now
SPECIFICATIONS
Environmentally Preferable