Adjustable Chrome Wire Shelving Systems for Storage, Omcan Scientific
Supplier: Omcan Scientific
20098 20099 20100 20101 20102 20103 20104 20105 20106 20107 20108 20109 20110 20111 20112 20113 24226 24227 24228 24229 24230 24231 24233 20114 20115 20116 20117 20118 20119 20120 20121
CA11012-304EA 40.67 CAD
CA11012-304 CA11012-306 CA11012-308 CA11012-310 CA11012-312 CA11012-314 CA11012-316 CA11012-318 CA11012-320 CA11012-322 CA11012-324 CA11012-326 CA11012-328 CA11012-330 CA11012-332 CA11012-334 CA11012-336 CA11012-338 CA11012-340 CA11012-342 CA11012-344 CA11012-346 CA11012-350 CA11012-352 CA11012-354 CA11012-356 CA11012-358 CA11012-360 CA11012-362 CA11012-364 CA11012-366
Adjustable Chrome Wire Shelving Systems for Storage, Omcan Scientific
Furniture Shelves
The Omcan storage solutions are ideal for keeping stock within reach. These chrome shelves are sturdy and welded to endure tough everyday handling.
  • Convert unused space into a neatly organized storage center adaptable to your specific needs
  • Open wire construction maximizes light and circulation of air to keep stored items dry and ventilated
Capacity: Shelves under 1219 mm (48") in length accommodate up to 386 kg (850 lbs.) per shelf, shelves over 48" in length, accommodate up to 272 kg (600 lbs.) per shelf.
Certifications: NSF