Bizwell Iona FR Coveralls, Portwest
Supplier: Portwest
UBIZ5ORRM UBIZ5ORRXL UBIZ5ORRXXL UBIZ5ORRXXXL UBIZ5NAR4XL UBIZ5NAR5XL UBIZ5NARL UBIZ5NARM UBIZ5NARS UBIZ5NARXL UBIZ5NARXXL UBIZ5NARXXXL UBIZ5NATL UBIZ5NATM UBIZ5NATXL UBIZ5NATXXL UBIZ5NATXXXL UBIZ5ORR4XL UBIZ5ORR5XL UBIZ5ORRL
76105-564EA 259.74 CAD
76105-564 76105-566 76105-568 76105-570 76105-644 76105-646 76105-648 76105-650 76105-652 76105-654 76105-656 76105-658 76105-660 76105-662 76105-664 76105-666 76105-668 76105-670 76105-672 76105-674
Bizwell Iona FR Coveralls, Portwest
Workwear
Coveralls are designed to protect against radiant, convective, and contact heat.
  • Pockets: two front, two back, one concealed cell phone pocket, two side, two knee, and one rule
  • Highly durable, flame resistant reflective tape
  • Dual hazard protection
  • ARC2
Certified protection against molten metal splash and guaranteed flame resistance for the life of the garment.
Certifications: EN ISO 11612 (A1+A2. B1. C1. E1. F1). EN ISO 11611 Class 1 (A1+A2). NFPA® 70E. ASTM F1506-10a. NFPA® 2112. ASTM F1959/F1959M-12 (ATPV 11.2 CAL/CM2 (HAF = 80.4%)).