BizTex Microporous Coveralls, Portwest
Supplier: Portwest
ST40WHRL ST40WHRM ST40WHRXL ST40WHRXXL
76105-494EA 481.05 CAD
76105-494 76105-496 76105-498 76105-596
BizTex Microporous Coveralls, Portwest
Disposable Garments
Excellent liquid and particulate barrier protection.
  • Durable and abrasion resistant
  • Elasticated hood, ankles, cuffs, and waist
  • Low linting construction
  • Anti-static
Coveralls feature a two-way front zipper and sealable front flap.
Certifications: EN 13034 Type 6B. EN ISO 13982 Type 5B. EN 1073-2. EN 14126. ANSI/ISEA 101. EN 1149 -5.