BizTex SMS Coveralls, Portwest
Supplier: Portwest
ST30WHRL ST30WHRM ST30WHRXL ST30WHRXXL ST30WHRXXXL
76105-484EA 775.67 CAD
76105-484 76105-486 76105-488 76105-490 76105-492
BizTex SMS Coveralls, Portwest
Disposable Garments
Coveralls provide an effective barrier against particulates, aerosols, and light sprays.
  • Highly breathable SMS fabric
  • Elasticated hood, ankles, cuffs, and waist
  • Anti-static
Coveralls feature a two-way front zipper and sealable front flap.
Certifications: EN ISO 13982 Type 5. EN 1073-2. ANSI/ISEA 101. EN 1149 -5. EN 13034 Type 6.