Hazmax® Hazardous Material Boots, Onguard
Supplier: Onguard Industries
87012-09 87012-10 87012-12 87012-14 87012-06 87012-07 87012-08 87012-11 87012-13 87012-15
21921-965EA 297.71 CAD
21921-965 21921-968 21921-972 21921-976 21921-960 21921-962 21921-964 21921-970 21921-974 21921-978
Hazmax® Hazardous Material Boots, Onguard
Shoes and Boots
The boot that continues to set the standard for biological hazards and waste protection.
  • Steel toe, steel shank, steel midsole with Ultragrip® sipe outsole
  • 16" boot height
  • Exceeds CSA standards for dielectric footwear
  • One-piece injection molded construction
  • Replaceable cushioned insoles
  • Cut off bands
  • Kick off lug
Hazmax® hazardous material boots are the finest boots made for hazardous material handling. They are manufactured from a high viscosity and flexible PVC alloy, and feature a non-absorbent polyester lining for easy cleaning and quick drying.More Product Information