Polyblend Steel Toe Workshoes with Cleated Outsole, Onguard
Supplier: Onguard Industries
86104-06 86104-07 86104-08 86104-09 86104-10 86104-11 86104-12 86104-13
21922-082EA 74.83 CAD
21922-082 21922-084 21922-086 21922-088 21922-090 21922-092 21922-094 21922-096
Polyblend Steel Toe Workshoes with Cleated Outsole, Onguard
Shoes and Boots
The Polyblend compound is designed to provide moderate chemical resistance.
  • Protective toe cap
  • Comfortable, waterproof design
  • Fuel oil resistant
  • Reinforced steel shank for added safety and comfort
  • Unique PVC/polyurethane formula provides moderate chemical resistance
  • Non-slip, self cleaning soles
  • Cleated outsole to prevent clogging on wet or muddy surfaces
Boots feature a protective toe cap that offers maximum protection against falling objects. The built-in protective toe cap serves to protect from impacts at an energy level of 125 J and a compressionMore Product Information