Plain Toe PVC Safety Boots with Steel Shank, Onguard
Supplier: Onguard Industries
81011-10 81011-09 81011-11 81011-12 81011-06 81011-07 81011-08
10466-476PR 72.89 CAD
10466-476 76047-212 76047-214 76047-216 76047-236 76047-238 76047-240
Plain Toe PVC Safety Boots with Steel Shank, Onguard
Shoes and Boots
Boots provide reliable, industry proven protection.
  • Comfortable, waterproof protection
  • Superior traction on slippery surfaces
  • PVC upper provides mild chemical resistance and helps comply with HACCP guidelines
Injection-molded seamless construction with replaceable cushion insoles. Boots feature a Safety-Loc™ sole, cut-off bands for height adjustment, steel shank, and pull-up/kick-off lugs for easy on/off. These boots offer a perfect fit, optimizing comfort for everyday use. They are 100% waterproof to keep feet dry under wet and dirty conditions, and offer protection against a wide variety of chemicals. Excellent for use in commercial fishing, shrimping, and food processing.