Sanitex Jackets with Hood Snaps, Onguard
Supplier: Onguard Industries
71232-SM 71232-MD 71232-LG 71232-XL 71232-2X 71232-3X 71232-4X 71232-5X
21922-206EA 100.22 CAD
21922-206 21922-208 21922-210 21922-212 21922-214 21922-216 21922-218 21922-220
Sanitex Jackets with Hood Snaps, Onguard
Workwear
Protection for all sanitation situations and needs.
  • Heavy-duty PVC on high count nylon polyester
  • Green finish
  • 0.35 mm material thickness
Specially designed for use in chemical plants, refineries, food processing, and environmental clean-up applications. Garments are manufactured from heavy-duty PVC on high count nylon polyester. Reinforced elbows to prevent splitting. Storm flap front and heavy-duty zipper. Cuff take-up snaps ensure a snug/tapered fit.
Certifications: Complies with ASTM D6413 for flame resistance.