LD-100 System Frocks, 3508/3509/3510 Series, WORKLON®
Supplier: Superior Uniform
3509-XS 3509-S 3509-M 3509-L 3509-XL 3509-2XL 3509-3XL 3509-4XL 3509-5XL 3508-XS 3508-S 3508-M 3508-L 3508-XL 3508-2XL 3508-3XL 3508-4XL 3508-5XL 3510-XS
89136-732EA 142.86 CAD
89136-732 89136-734 89136-736 89136-738 89136-740 89136-742 89136-744 89136-746 89136-748 89136-750 89136-752 89136-754 89136-756 89136-758 89136-760 89136-762 89136-764 89136-766 89136-768
LD-100 System Frocks, 3508/3509/3510 Series, WORKLON®
Workwear
Snap closure, raglan sleeve frocks feature polyester knit cuffs and two-ply reinforced placket.
  • Adjustable snaps at collar
  • Color-coded hang loop denoting size
  • Unisex design in a wide range of sizes
Garments are manufactured from 99% polyester, 1% carbon yarn Maxima® high density ESD polyester with a carbon yarn stripe. This fabric can be used in a variety of industries including pharmaceutical, biotechnical, biological, and food processing. Ideal for applications where particulate control, and fluid and bacteria resistance is essential.
Certifications: Engineered to meet up to Class 10 (ISO 4) cleanroom requirements.