HD-10 System Hypalon Sole Shoe Covers, WORKLON®
Supplier: Superior Uniform
1207-XS 1207-S 1207-M 1207-L 1207-XL 1207-2XL
89136-786PR 65.76 CAD
89136-786 89136-788 89136-790 89136-792 89136-794 89136-796
HD-10 System Hypalon Sole Shoe Covers, WORKLON®
Workwear
Full elastic opening and rear snap adjustment.
  • Generous sizing
  • Color-coded stitching denoting size
  • Unisex design in a wide range of sizes
Densely woven, 100% polyester fabric offers excellent barrier protection against bacteria, fluids, and particles. These shoe covers can be worn in a variety of industries.
Certifications: Designed for cleanrooms up to and including Class 10 (ISO 4), where bacteria control is essential.
Order Now
SPECIFICATIONS
Model HD-10
Environmentally Preferable
Garment application Industrial,Laboratory
Risk area
Gender Unisex
Closure type Press studs / snaps
Fabric 100% Polyester