Sterility Maintenance Covers, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
815 820 825 830 835 840 845 865 855SMC 880SMC
76358-962CS 453.37 CAD
76358-962 76358-964 76358-966 76358-968 76358-970 76358-972 76358-974 76358-976 76358-978 76358-980 76358-982 76359-148 76359-150
Sterility Maintenance Covers, Medegen Medical Products
Covers
Sterility maintenance covers provide optimal protection of processed medical equipment and stored supplies.
  • Wide range of sizes
  • Available with self seal adhesive or heat seal closure
  • Easy opening linear tear strip allows for quick access to contents
  • Tamper evident
Ideal for use in areas where sterile products are packaged or reprocessed.