Anti-Static Kick Bucket Liners, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
50-01 50-02 50-04
76359-120CS 153.77 CAD
76359-120 76359-122 76359-124
Anti-Static Kick Bucket Liners, Medegen Medical Products
Bags Waste Disposal Bags
Anti-static kick bucket liners comply with operating room requirements.
  • Bags with size of 12×24×8" fit round kick buckets
  • Bags with size of 18×16×8" fit oval kick buckets
  • Red film bags feature a biohazard print
  • Blue film bags feature a soiled linen print
Liners contain a carbon additive to help prevent the build-up of electrostatic charges.
Certifications: Meets NFPA Standard 56A.