R10000 M611, Printer Ribbons for BMP®61 Labeling System BradyPrinter™, Brady
Supplier: Brady Worldwide
M61-R10010 M61-R10010-BL M61-R10010-GN M61-R10010-RD M61-R10010-WT M61-R10010-YL
76359-380RL 200.16 CAD
76359-380 76359-382 76359-384 76359-386 76359-388 76359-390 76359-392
R10000 M611, Printer Ribbons for BMP®61 Labeling System BradyPrinter™, Brady
Labels
Switch out ribbons in seconds with these easy-to-load ribbon cartridges that slide into place in the always-correct direction.
  • Use with BradyPrinter™ M611 and BMP® 61 labeling system
  • Single color ribbon
Durable R10000 series ribbons are based on a resin formulation that produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions. These self-contained ribbon cartridges are easy to replace, eliminating the hassle of printer calibration and ribbon threading. These ribbons are ideal for use with long-lasting indoor/outdoor materials such as Brady B-595.