Kimble® Silicone Lids, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
291111111 291111128 291111136 291112116 291112124 291112132 291113112 291113129 291113137 291110012 291110029 291110037
76360-016EA 3.57 CAD
76360-016 76360-018 76360-020 76360-022 76360-024 76360-026 76360-028 76360-030 76360-032 76360-034 76360-036 76360-038
Kimble® Silicone Lids, DWK Life Sciences
Closures Bottle Closures
The reusable KIMBLE® silicone lids stretch easily and adapt to many container sizes, improves lab safety, enhances sample management, offers a more sustainable and cost-effective solution over foils, films and other single-use covers.
  • Versatile, safe, easily identifiable, and sustainable
  • They are suitable for a wide range of applications, and can be used to cover and seal a variety of differently shaped lab containers
  • Lids seal flush with the container and protect contents from dust or splashes
  • With the help of the integrated pull tabs, the lid can be easily removed by hand or with gloves
  • Three different colors (pink, cyan, and green) make it easy to color mark different samples
  • Sample information can be written with a lab marker on the marking field flap for quick identification
In order to prevent contamination during reactions in the laboratory environment, safety cover has been developed: The innovative Kimble® silicone lid. Due to its excellent stretchability, the lid can be used as a cover for a wide variety of containers of different diameters and shapes. The wide overlapping sides of the silicone lid grips the vessel wall. This is useful, for example during mixing, to avoid leakage or splashing of the reaction medium. Because of the nature of the material, these silicone lids are easy to clean, reusable, and offer an environmentally friendly and economical alternative to other types of covers and films.