Cyanine 750 succinimidyl ester cyanine dye
Supplier: Biotium
90049-1MG 90049-10MG
10098-678EA 352.45 CAD
10098-678 10098-680
Cyanine 750 succinimidyl ester cyanine dye
Cyanine succinimidyl ester
Biotium offers high quality amine-reactive succinimidyl ester (SE) dyes used to label proteins, oligonucleotides, and nucleic acids.
  • Cyanine 750 is structurally identical to Cy®7 from GE Healthcare.
  • λEx/λEm = 755/777 nm
  • Dark blue solid soluble in DMF and DMSO
  • Store at -20°C and protect from moisture and light
We offer Cyanine 555 (Cy®3), Cyanine 647 (Cy®5), Cyanine 680 (Cy®5.5), and Cyanine 750 (Cy®7) as high quality succinimidyl esters.
Also see our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy3, Cy5, Cy5.5, and Cy7 are registered trademarks of GE Healthcare.