BRAND® Sealing Film 8-Strips for PCR and Storage, BrandTech Scientific
Supplier: Brandtech
781382 781383
76319-826PK 138.06 CAD
76319-826 76319-828
BRAND® Sealing Film 8-Strips for PCR and Storage, BrandTech Scientific
Seals Microplate Seals
An excellent choice for sealing PCR 8-strips, or sealing/re-sealing partial plates.
  • 8-Strips of polyester or aluminum
  • DNA-, RNase- and DNase-free
The sheets contain 6 to 8 perforated strips for convenience.