VITLAB® Erlenmeyer Flasks, PP, with GL 45 Screw Cap, BrandTech Scientific
Supplier: Brandtech
566941 567941 568941 569941
76313-834PK 102.92 CAD
76313-834 76313-836 76313-838 76313-840 76313-886
VITLAB® Erlenmeyer Flasks, PP, with GL 45 Screw Cap, BrandTech Scientific
Flasks Erlenmeyer Flasks
These flasks have tight fitting screw caps, and feature resistant screened graduations.
  • Made of chemical- and break-resistant PP
  • Blue screened print for legibility
All sizes have GL 45 threads and interchangeable caps.