Polymix® Laboratory Grinding Mills, PX-MFC 90 D, Kinematica
Supplier: Kinematica
POLYMIX®
35010033 35010023
76359-404EA 19850.3 CAD
76359-404 76359-402
Polymix® Laboratory Grinding Mills, PX-MFC 90 D, Kinematica
Mills
Grinding mill with powerful motor, which is suitable for all laboratory dry milling applications.
  • Powerful motor provides speeds up to 6000 min-1 with low noise emissions (less than 70 dB(A) at 6000 min-1 without load) due to brushless 3 phase AC motor
  • Milling chamber can be equipped with hammer grinding or blade grinding attachment (second milling chamber not required)
  • Integrated speed controller with LED display shows speed and status messages
  • Integrated overheating, overload and blocking protection means motor shuts down if overload conditions arise, ensuring the motor is protected
  • Safety switch for automatic switch-off, and energy saving automatic standby mode
Milling chamber with attached funnel and rotary slide for sample feed, can be used with interchangeable hammer or blade grinding attachments, and sieves with mesh sizes from 0.2 up to 6 mm. Chamber parts in contact with the sample are made from hardened stainless steel, the anodized aluminum base frame has an integrated handle. The ground material can be collected using vessels with NS 29/32 standard connections (holding clip necessary) or receiving tubes.
This laboratory mill is characterized by a very robust design using solid milled parts and hardened components that have been designed for high wear and tear found with such processes as grinding of stones which can have a hardness of up to 6 on the Mohs scale. You will receive tried and tested Swiss quality.