VWR® Cryo Racks for 1.0, 1.8 and 4.5 ml Cryogenic Tubes, PC
Supplier: VWR
VWR®
5700-19-22AV1 5730-19-22AV1 5750-19-22AV1 5720-19-22AV1
76337-368CS 653.32 CAD
76337-368 76337-364 76337-366 76337-362
VWR® Cryo Racks for 1.0, 1.8 and 4.5 ml Cryogenic Tubes, PC
Racks Cryogenic Vial Racks
These racks are ideal for long-term cryogenic storage.
  • Drains and vents prevent pressure build-up
  • Crystal-clear lid provides sample visibility
  • Alphanumeric indexing for greater organization
Maximum tube dimensions:
12×49 mm tubes fit: 76337-368, 76337-366 and 76337-362
12×90 mm tubes fit: 76337-364