VWR® Microtube Racks, 64-Place
Supplier: VWR International
76333-282 76333-294
76333-282CS 755.21 CAD
76333-282 76333-294
VWR® Microtube Racks, 64-Place
Racks Tube Racks
The 64-place freezer storage rack is designed to replace 51/4×51/4" cardboard freezer storage boxes.
  • Rack holds up to 64 tubes in an 8×8 matrix
  • Produced from copolymer polypropylene
  • Withstands limited autoclaving (up to 15 minutes) as well as deep freezer storage (down to –90 °C)
  • Both sides are labeled with a large easy-to-read 1 to 8 by A-H matrix
Designed to be reversible; one side has holes suitable for the larger tubes the other has holes suitable for smaller size microtubes. The hole spacing is greater than with the standard 80-place rack makingMore Product Information