VWR® Microtube Racks, 96-Place
Supplier: VWR International
76333-304 76333-306 76333-308 76333-310 76333-312 76333-314 76337-380 76337-382
76333-304CS 315.86 CAD
76333-304 76333-306 76333-308 76333-310 76333-312 76333-314 76337-380 76337-382
VWR® Microtube Racks, 96-Place
Racks Tube Racks
Double-sided racks hold 1.7 and 2.0 ml tubes on one side and 0.5 ml tubes on the reverse side.
  • Rack for use with 0.5, 1.7, and 2.0 ml microcentrifuge tubes and 2.0 ml cryogenic storage vials
  • Rack and clear cover (sold separately) are autoclavable and freezable
Racks are available in eight colors and an assorted color pack with one of each color: blue, green, orange, red, and yellow.
Ordering information: Clear cover sold separately.