Urinary Drainage Bags, Medegen Medical Products
Supplier: Medegen Medical Products
2556 2550 2555
76329-594CS 578.15 CAD
76329-594 76329-598 76329-596
Urinary Drainage Bags, Medegen Medical Products
Bags Sample Bags
These urinary drainage bags have sterile fluid pathways and anti-reflux valves to prevent urinary backflow to help reduce risk of infection.
  • All bags are designed with built in handles for easy attachments to beds, carts, wheelchairs, walkers and more
  • Additional clip is provided for securing bags to clothing
Leg bags come with adjustable latex strap and are available in M and L sizes.
Graduated in 200 cc's (ml) increments upto 2000 cc (ml), drainage bags come with a latex sampling port for aeseptic retrival. Graduated in 100 cc and 4 oz. increments. Medium bags are calibrated to 600 cc (ml) and large to 800 cc (ml).
Delivery information: Available in standard or leg drainage bag styles.
Packaging: Packed in polybags.