Spouted Collection Containers, Medegen Medical Products
Supplier: Medegen Medical Products
4647 M4630 4629 M4646
76329-650CS 253.16 CAD
76329-650 76329-654 76329-652 76329-656
Spouted Collection Containers, Medegen Medical Products
Sample Containers General Purpose Containers
These 8 oz. spouted containers are available in both polystyrene and polypropylene. Both options come with a convenient pour spout for simplified specimen transfer.
  • Graduated in 0.5 oz. and 10 cc (ml) increments
  • Polystyrene provides crystal clear clarity
  • Polypropylene (translucent) provides improved durability
Ordering information: Printed lids can be purchased separately.