ICERoam35 Mobile Dry Ice Reusable Transport Container
Supplier: ROEMER INDUSTRIES, INC.
ICEROAM35
76333-166EA 15141.95 CAD
76333-166
ICERoam35 Mobile Dry Ice Reusable Transport Container
Coolers Dry Ice Chests
Withstand over 200 trips by air, land and sea with military-grade aluminum.
  • Over 3" of injected polyurethane insulation
  • 35 liters of storage space
  • Water tight, TIG welded aluminum payload compartment tub
With dry ice in container, temperature will hold for three to five days.
Refrigerated Payload Dimension: (L × W × H) 19×17×7"