Chromatography Reservoirs, Rodaviss Joint, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0414250 CS-C0414500 CS-C0414000 CS-C0415500 CS-C041451L CS-C041452L CS-C0414100
CA10201-258CS 245.36 CAD
CA10201-258 CA10201-260 CA10201-262 CA10201-264 CA10201-266 CA10201-268 CA10204-190
Chromatography Reservoirs, Rodaviss Joint, ChemScience
Reagent Reservoirs
These reservoirs have a top 24/40 or 45/50 Rodaviss outer joint for use with flow controller, and the listed size lower inner joint.
Delivery information: Supplied complete with open top cap, Viton O-ring and loosening ring.