Chromatography Columns, Rodaviss Joint, Fritted Disc, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0404172C CS-C0404173C CS-C0404174C CS-C0404261C CS-C0404322C CS-C0404323C CS-C0404324C CS-C0404461C CS-C0405801C CS-C0405802C CS-C0405803C CS-C0405804C CS-C0404171C CS-C0404262C CS-C0404263C CS-C0404264C CS-C0404321C CS-C0404462C CS-C0404463C CS-C0404464C CS-C0404601C CS-C0404602C CS-C0404603C CS-C0404604C CS-C0404605C CS-C0405805C
CA10201-300CS 510.15 CAD
CA10201-300 CA10201-302 CA10201-304 CA10201-306 CA10201-308 CA10201-310 CA10201-312 CA10201-314 CA10201-316 CA10201-318 CA10201-320 CA10201-322 CA10203-606 CA10203-608 CA10203-610 CA10203-612 CA10203-614 CA10203-616 CA10203-618 CA10203-620 CA10203-622 CA10203-624 CA10203-626 CA10203-628 CA10203-630 CA10203-632
Chromatography Columns, Rodaviss Joint, Fritted Disc, ChemScience
Chromatography Columns
These columns have a Teflon® stopcock, a Rodaviss joint and a fritted disc.
  • Equipped with a coarse porosity sealed in fritted disc to support column packing
The unique design of the Rodaviss joint affords a grease free seal while holding glassware together without clamps.
Delivery information: Columns are supplied complete with open top cap loosening ring and O-ring.