Chromatography Columns, Rodaviss Joint, ChemScience
Supplier: SATI International Scientific
CS-C0404605 CS-C0405802 CS-C0404602 CS-C0404463 CS-C0404601 CS-C0404173 CS-C0404462 CS-C0404261 CS-C0405801 CS-C0404322 CS-C0404174 CS-C0404604 CS-C0405805 CS-C0405804 CS-C0404464 CS-C0404172 CS-C0404321 CS-C0404603 CS-C0404171 CS-C0404263 CS-C0404262 CS-C0404264 CS-C0404324 CS-C0404323 CS-C0405803 CS-C0404461
CA10201-288CS 565.15 CAD
CA10201-288 CA10203-590 CA10201-286 CA10203-598 CA10201-280 CA10203-596 CA10203-594 CA10201-284 CA10203-592 CA10201-282 CA10203-604 CA10203-602 CA10203-588 CA10203-600 CA10201-298 CA10201-276 CA10201-274 CA10201-296 CA10201-278 CA10203-586 CA10201-290 CA10203-584 CA10201-272 CA10201-294 CA10201-270 CA10201-292
Chromatography Columns, Rodaviss Joint, ChemScience
Chromatography Columns
These columns have a Teflon® stopcock and a Rodaviss joint.
The unique design of the Rodaviss joint affords a grease free seal while holding glassware together without clamps.
Delivery information: Columns are supplied complete with open top cap loosening ring and O-ring.