ICP Calibration Standard 15, VeriSpec®
Supplier: Ricca Chemical
RV010672-100N
RV010672-500N
RCRV010672100NEA
846.88
CAD
RCRV010672100N
RCRV010672500N
ICP Calibration Standard 15, VeriSpec®
Standards
Cationic Standards
Multi-Element Standards
15-component standard in 5% nitric acid
5 ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se Sr, Zn; 50 ppm K
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility