Chromatography Columns, With Top Outer Joint, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-F0386020C CS-F0386020M CS-F0386030C CS-F0386030M CS-F0386040C CS-F0386040M CS-F0388520C CS-F0388520M CS-F0388530C CS-F0388530M CS-F0388540C CS-F0384020C CS-F0384020M CS-F0384030C CS-F0384030M CS-F0388540M
CA10201-236CS 427.82 CAD
CA10201-236 CA10201-238 CA10201-240 CA10201-242 CA10201-244 CA10201-246 CA10201-248 CA10201-250 CA10201-252 CA10201-254 CA10201-256 CA10203-576 CA10203-578 CA10203-580 CA10203-582 CA10204-184
Chromatography Columns, With Top Outer Joint, ChemScience
Chromatography Columns
These short columns with lower 24/40 joint and hose connection to apply vacuum.
Column length is measured from bottom of top joint to fritted disc.