Chromatography Columns, With Reservoir, Spherical Joint, Fritted Disc, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0381730C CS-C0381740C CS-C0382620C CS-C0382625C CS-C0382630C CS-C0382640C CS-C0383220C CS-C0383225C CS-C0383230C CS-C0383240C CS-C0384620C CS-C0384625C CS-C0384630C CS-C0384640C CS-C0386020C CS-C0386025C CS-C0386030C CS-C0386040C CS-C0386060C CS-C0388020C CS-C0388025C CS-C0388030C CS-C0388040C CS-C0388060C
CA10201-164CS 410.79 CAD
CA10201-164 CA10201-166 CA10201-168 CA10201-170 CA10201-172 CA10201-174 CA10201-176 CA10201-178 CA10201-180 CA10201-182 CA10201-184 CA10201-186 CA10201-188 CA10201-190 CA10201-192 CA10201-194 CA10201-196 CA10201-198 CA10201-200 CA10201-202 CA10201-204 CA10201-206 CA10201-208 CA10201-210
Chromatography Columns, With Reservoir, Spherical Joint, Fritted Disc, ChemScience
Chromatography Columns
These columns with reservoir have a Teflon® stopcock and a 35/20 spherical joint.