Chromatography Columns, Spherical Joint, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0195171 CS-C0195172 CS-C0195173 CS-C0195261 CS-C0195262 CS-C0195263 CS-C0195264 CS-C0195321 CS-C0195322 CS-C0195323 CS-C0195324 CS-C0195461 CS-C0195462 CS-C0195463 CS-C0195464 CS-C0195601 CS-C0195602 CS-C0195603 CS-C0195604 CS-C0195605 CS-C0195801 CS-C0195802 CS-C0195803 CS-C0195804 CS-C0195805
CA10201-044CS 283.63 CAD
CA10201-044 CA10201-046 CA10201-048 CA10201-050 CA10201-052 CA10201-054 CA10201-056 CA10201-058 CA10201-060 CA10201-062 CA10201-064 CA10201-066 CA10201-068 CA10201-070 CA10201-072 CA10201-074 CA10201-076 CA10201-078 CA10201-080 CA10201-082 CA10201-084 CA10201-086 CA10201-088 CA10201-090 CA10203-552
Chromatography Columns, Spherical Joint, ChemScience
Chromatography Columns
These columns have a Teflon® stopcock and a spherical joint.
The spherical joint used with pinch clamp makes quick and tight connection under pressure for flash chromatography.
Order Now
SPECIFICATIONS
Environmentally Preferable