Chromatography Columns, With Reservoir, Standard Taper, Fritted Disc, ChemScience
Supplier: SATI International Scientific
CS-C0376040C CS-C0372640C CS-C0378060C CS-C0372630C CS-C0373230C CS-C0371720C CS-C0374625C CS-C0378030C CS-C0374630C CS-C0371730C CS-C0376060C CS-C0374640C CS-C0373220C CS-C0371740C CS-C0376020C CS-C0376025C CS-C0378040C CS-C0378025C CS-C0376030C CS-C0378020C CS-C0372625C CS-C0373240C CS-C0372620C CS-C0374620C CS-C0373225C
CA10201-024CS 486.48 CAD
CA10201-024 CA10201-002 CA10201-022 CA10201-000 CA10201-006 CA10201-028 CA10201-004 CA10201-026 CA10200-998 CA10201-042 CA10201-020 CA10201-040 CA10201-018 CA10201-012 CA10201-034 CA10201-032 CA10201-010 CA10201-038 CA10201-016 CA10201-036 CA10201-014 CA10201-030 CA10200-994 CA10200-996 CA10201-008
Chromatography Columns, With Reservoir, Standard Taper, Fritted Disc, ChemScience
Chromatography Columns
These columns with reservoir have a Teflon® stopcock and a 24/40 top outer joint.
They are equipped with a coarse porosity sealed in fritted disc to support column packing.