Schlenk Tubes, Modified, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-F0901410 CS-F0901425 CS-F0904200 CS-F0904500 CS-F0904000 CS-F0901410G CS-F0901425G CS-F0901450G CS-F0901950G CS-F0902450G CS-F0901100G CS-F0909100G CS-F0904100G CS-F0901200G CS-F0909200G CS-F0904200H CS-F0904500H CS-F0904000H CS-F0904500G CS-F0904000G CS-F0901425H CS-F0901450H CS-F0901950H CS-F0902450H CS-F0901100H CS-F0909100H CS-F0904100H CS-F0901200H CS-F0909200H CS-F0901450 CS-F0901950 CS-F0902450 CS-F0901100 CS-F0909100 CS-F0904100 CS-F0901200 CS-F0909200 CS-F0904200G
CA10202-270CS 146.72 CAD
CA10202-270 CA10202-272 CA10202-274 CA10202-276 CA10202-278 CA10202-280 CA10202-282 CA10202-284 CA10202-286 CA10202-288 CA10202-290 CA10202-292 CA10202-294 CA10202-296 CA10202-298 CA10202-300 CA10202-302 CA10202-304 CA10203-978 CA10203-980 CA10203-982 CA10203-984 CA10203-986 CA10203-988 CA10203-990 CA10203-992 CA10203-994 CA10203-996 CA10203-998 CA10204-440 CA10204-442 CA10204-444 CA10204-446 CA10204-448 CA10204-450 CA10204-452 CA10204-454 CA10204-590
Schlenk Tubes, Modified, ChemScience
Flasks Nitrogen Flasks
Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.