Schlenk Tubes, with High Vacuum Valve, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-F0580025 CS-F0580825 CS-F0580050 CS-F0580850 CS-F0580100 CS-F0584250 CS-F0588250 CS-F0584350 CS-F0588350
CA10202-250CS 187.24 CAD
CA10202-250 CA10202-252 CA10202-254 CA10202-256 CA10202-258 CA10202-260 CA10202-262 CA10202-264 CA10202-266 CA10202-268
Schlenk Tubes, with High Vacuum Valve, ChemScience
Flasks Nitrogen Flasks
These valves have a PTFE wiper to protect Viton® O-ring on shaft.
Delivery information: Supplied with high vacuum valve with PTFE O-ring at tip.
Order Now
SPECIFICATIONS
Environmentally Preferable