Chromatography Columns, With Reservoir, Standard Taper, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0371720 CS-C0371730 CS-C0372640 CS-C0373220 CS-C0373225 CS-C0373230 CS-C0373240 CS-C0374620 CS-C0374625 CS-C0374630 CS-C0374640 CS-C0376020 CS-C0376025 CS-C0378025 CS-C0378030 CS-C0378040 CS-C0378060 CS-C0371740 CS-C0372620 CS-C0372625 CS-C0372630 CS-C0376030 CS-C0376040 CS-C0376060 CS-C0378020
CA10200-960CS 273.95 CAD
CA10200-960 CA10200-962 CA10200-964 CA10200-966 CA10200-968 CA10200-970 CA10200-972 CA10200-974 CA10200-976 CA10200-978 CA10200-980 CA10200-982 CA10200-984 CA10200-986 CA10200-988 CA10200-990 CA10200-992 CA10203-544 CA10203-546 CA10203-548 CA10203-550 CA10204-160 CA10204-162 CA10204-164 CA10204-166
Chromatography Columns, With Reservoir, Standard Taper, ChemScience
Chromatography Columns
These columns with reservoir have a Teflon® stopcock and a 24/40 top outer joint.