Chromatography Columns, General Purpose, Standard Taper, Fritted Disc, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0184171C CS-C0184172C CS-C0184173C CS-C0184174C CS-C0184261C CS-C0184262C CS-C0184263C CS-C0184324C CS-C0184461C CS-C0184462C CS-C0184463C CS-C0184464C CS-C0184601C CS-C0184602C CS-C0184603C CS-C0184604C CS-C0184605C CS-C0184801C CS-C0184802C CS-C0184803C CS-C0184804C CS-C0184805C CS-C0184264C CS-C0184321C CS-C0184322C CS-C0184323C
CA10200-916CS 251.03 CAD
CA10200-916 CA10200-918 CA10200-920 CA10200-922 CA10200-924 CA10200-926 CA10200-928 CA10200-930 CA10200-932 CA10200-934 CA10200-936 CA10200-938 CA10200-940 CA10200-942 CA10200-944 CA10200-946 CA10200-948 CA10200-950 CA10200-952 CA10200-954 CA10200-956 CA10200-958 CA10204-152 CA10204-154 CA10204-156 CA10204-158
Chromatography Columns, General Purpose, Standard Taper, Fritted Disc, ChemScience
Chromatography Columns
These general-purpose columns have a Teflon® stopcock and a top outer joint.
They are equipped with a coarse porosity fritted disc to support column packing.