VeriSpec™ Zirconium Standards, for ICP, RICCA Chemical Company
Supplier: Ricca Chemical
RV010331-100N RV010331-250N RV010331-500N RV010432-100N RV010501-100N
RCRV010331100NEA 205.4 CAD
RCRV010331100N RCRV010331250N RCRV010331500N RCRV010432100N RCRV010501100N
VeriSpec™ Zirconium Standards, for ICP, RICCA Chemical Company
Standards Cationic Standards Zirconium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Order Now
SPECIFICATIONS
Environmentally Preferable