VeriSpec™ Zinc Standards, for ICP, RICCA Chemical Company
Supplier: Ricca Chemical
RV010431-100N
RV010500-100N
RV010500-500N
RCRV010431100NEA
377.62
CAD
RCRV010431100N
RCRV010500100N
RCRV010500500N
VeriSpec™ Zinc Standards, for ICP, RICCA Chemical Company
Standards
Cationic Standards
Zinc Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml