Yttrium Standard, for ICP, VeriSpec®, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010429-100N RV010429-250N RV010429-500N
RCRV010429100NEA 408.64 CAD
RCRV010429500N RCRV010429100N RCRV010429250N
Yttrium Standard, for ICP, VeriSpec®, RICCA Chemical Company
Standards Cationic Standards Yttrium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml