Triethanolamine - HN(C₂H₄OH)₃+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010948-100N RV010948-250N RV010948-500N
RCRV010948100NEA 188.02 CAD
RCRV010948100N RCRV010948250N RCRV010948500N
Triethanolamine - HN(C₂H₄OH)₃+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Triethanolammonium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml