Titanium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010322-100N RV010322-250N RV010322-500N RV010358-100N RV010358-250N RV010358-500N RV010423-100N RV010492-100N
RCRV010322100NEA 189.6 CAD
RCRV010322100N RCRV010322250N RCRV010322500N RCRV010358100N RCRV010358250N RCRV010358500N RCRV010423100N RCRV010492100N
Titanium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Titanium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml