Titanium Standard, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010056-100N RV010056-500N
RCRV010056100NEA 112.18 CAD
RCRV010056100N RCRV010056500N
Titanium Standard, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Titanium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml