Terbium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010420-250N RV010420-100N RV010420-500N
RCRV010420100NEA 676.48 CAD
RCRV010420250N RCRV010420500N RCRV010420100N
Terbium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Terbium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml