Tantalum Standard, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010054-100N
RV010054-500N
RCRV010054100NEA
126.4
CAD
RCRV010054100N
RCRV010054500N
Tantalum Standard, for AAS, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Tantalum Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml