VeriSpec® Silver Standard for ICP/MS in 2% HNO₃ Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility, Ricca Chemical Company
Supplier: Ricca Chemical
RV010502-100N RV010571-100N
RCRV010502100NEA 241.74 CAD
RCRV010502100N RCRV010571100N
VeriSpec® Silver Standard for ICP/MS in 2% HNO₃ Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility, Ricca Chemical Company
Standards Cationic Standards Silver Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml