Ruthenium Standards, for ICP/MS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010548-100N
RV010616-100N
RCRV010548100NEA
298.62
CAD
RCRV010548100N
RCRV010616-50N
RCRV010616100N
Ruthenium Standards, for ICP/MS, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Ruthenium Standards
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml