Rhodium Standards, for ICP/MS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010547-100N RV010615-100N
RCRV010547100NEA 741.02 CAD
RCRV010547100N RCRV010615-50N RCRV010615100N
Rhodium Standards, for ICP/MS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Rhodium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml